
Plastic additive 2

Synonym(s):
Pentaerythritol tetrakis(3,5-di-tert-butyl-4-hydroxyhydrocinnamate), Pentaerythritol tetrakis(3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate)
Linear Formula:
[HOC6H2[C(CH3)3]2CH2CH2CO2CH2]4C
CAS Number:
Molecular Weight:
1177.63
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Showing 1-2 of 2