
Iodixanol impurity C

Synonym(s):
5-[Acetyl[3-[[3,5-bis[(2,3-dihydroxypropyl)carbamoyl]-2,4,6-triiodophenyl]amino]-2-hydroxypropyl]amino]-N,N′-bis(2,3-dihydroxypropyl)-2,4,6-triiodobenzene-1,3-dicarboxamide
Empirical Formula (Hill Notation):
C33H42I6N6O14
CAS Number:
Molecular Weight:
1508.15
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing