
Econazole

Empirical Formula (Hill Notation):
C18H15Cl3N2O
CAS Number:
Molecular Weight:
381.68
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing