
DPPF ChemBeads

Synonym(s):
DPPF, 1,1′-Ferrocenediyl-bis(diphenylphosphine), dppf
Empirical Formula (Hill Notation):
C34H28FeP2
CAS Number:
Molecular Weight:
554.38
Products
Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing