
cis,cis-1,3,5-Trimethylcyclohexane-1,3,5-tricarboxylic acid

Synonym(s):
Kemp’s triacid
Empirical Formula (Hill Notation):
C12H18O6
CAS Number:
Molecular Weight:
258.27
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing