
Ciprofloxacin Impurity B (Ph. Eur.)

Synonym(s):
1-Cyclopropyl-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid, Desfluorociprofloxacin
Empirical Formula (Hill Notation):
C17H19N3O3
CAS Number:
Molecular Weight:
313.35
Products
Brand
Formula Weight
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing