
Bromocriptine mesilate for system suitability

Synonym(s):
2-Bromo-α-ergocryptine methanesulfonate salt, (+)-2-Bromo-12′-hydroxy-2′-(1-methylethyl)-5′-(2-methylpropyl)ergotaman-3′,6′-18-trione methanesulfonate salt, (+)-Bromocriptine methanesulfonate salt, Bromocriptine mesylate salt
Empirical Formula (Hill Notation):
C32H40BrN5O5 · CH4SO3
CAS Number:
Molecular Weight:
750.70
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing