
Afzelin

Synonym(s):
3,4′,5,7-Tetrahydroxyflavone 3-rhamnoside, Afzeloside, Kaempferin, Kaempferol 3-O-alpha-L-rhamnoside, Kaempferol 3-rhamnoside
Empirical Formula (Hill Notation):
C21H20O10
CAS Number:
Molecular Weight:
432.38
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing