
(2-Amino-5-nitrophenyl)(2-chlorophenyl)methanone

Synonym(s):
2-Amino-2′-chloro-5-nitrobenzophenone, (2-Amino-5-nitrophenyl)(2-chlorophenyl)methanone
Empirical Formula (Hill Notation):
C13H9ClN2O3
CAS Number:
Molecular Weight:
276.68
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing