Merck
  • Visible light initiated release of calcium ions through photochemical electron transfer reactions.
Visible light initiated release of calcium ions through photochemical electron transfer reactions.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2017-05-13)
Romina R Heymann, Matthew D Thum, Apryl L Hardee, Daniel E Falvey
ABSTRACT
Photolysis of anthraquinone or flavin photosensitizers in the presence of calcium EDTA complexes results in decomposition of the EDTA complex, releasing free Ca
Sigma-Aldrich
Sodium anthraquinone-2-sulfonate, ≥98% (HPLC)
Pricing and availability is not currently available.