Merck
  • High potassium promotes mutual interaction between (pro)renin receptor and the local renin-angiotensin-aldosterone system in rat inner medullary collecting duct cells.
High potassium promotes mutual interaction between (pro)renin receptor and the local renin-angiotensin-aldosterone system in rat inner medullary collecting duct cells.
American journal of physiology. Cell physiology (2016-08-19)
Chuanming Xu, Hui Fang, Li Zhou, Aihua Lu, Tianxin Yang
ABSTRACT
(Pro)renin receptor (PRR) is predominantly expressed in the collecting duct (CD) with unclear functional implication. It is not known whether CD PRR is regulated by high potassium (HK). Here, we aimed to investigate the effect of HK on PRR expression and its role in regulation of aldosterone synthesis and release in the CD. In primary rat inner medullary CD cells, HK augmented PRR expression and soluble PPR (sPRR) release in a time- and dose-dependent manner, which was attenuated by PRR small interfering RNA (siRNA), eplerenone, and losartan. HK upregulated aldosterone release in parallel with an increase of CYP11B2 (cytochrome P-450, family 11, subfamily B, polypeptide 2) protein expression and upregulation of medium renin activity, both of which were attenuated by a PRR antagonist PRO20, PRR siRNA, eplerenone, and losartan. Similarly, prorenin upregulated aldosterone release and CYP11B2 expression, both of which were attenuated by PRR siRNA. Interestingly, a recombinant sPRR (sPRR-His) also stimulated aldosterone release and CYP11B2 expression. Taken together, we conclude that HK enhances a local renin-angiotensin-aldosterone system (RAAS), leading to increased PRR expression, which in turn amplifies the response of the RAAS, ultimately contributing to heightened aldosterone release.
Sigma-Aldrich
Anti-Cytochrome P450 11β-Hydroxylase Antibody, clone 11B2-1A4, culture supernatant, clone 11B2-1A4, Chemicon®