Merck
  • Identification of sumoylation sites in CCDC6, the first identified RET partner gene in papillary thyroid carcinoma, uncovers a mode of regulating CCDC6 function on CREB1 transcriptional activity.
Identification of sumoylation sites in CCDC6, the first identified RET partner gene in papillary thyroid carcinoma, uncovers a mode of regulating CCDC6 function on CREB1 transcriptional activity.
PloS one (2012-11-13)
Chiara Luise, Francesco Merolla, Vincenza Leone, Simona Paladino, Daniela Sarnataro, Alfredo Fusco, Angela Celetti
ABSTRACT
CCDC6 was originally identified in chimeric genes as caused by chromosomal translocation involving the RET protooncogene in some thyroid tumors. Recognised as a 65 kDa pro-apoptotic phosphoprotein, CCDC6 has been enrolled as an ATM substrate that contribute to protect genome integrity by modulating PP4c activity in response to genotoxic stress. Recently, CCDC6 has been identified as a repressor of CREB1-dependent transcription. Sumoylation has emerged as an important mechanism in transcriptional control. Here, we report the identification and characterization of three sites of sumoylation in CCDC6 (K74, K266 and K424) which are highly conserved in vertebrates. We demonstrate that the post-translational modifications by SUMO2 constrain most of the CCDC6 protein in the cytosol and affect its functional interaction with CREB1 with a decrease of CCDC6 repressive function on CREB1 transcriptional activity. Indeed, the impairment of functional outcome of sumoylated CCDC6 is obtained knocking down all three the sumoylation sites. Interestingly, in thyroid cells the SUMO2-mediated CCDC6 post-translational modifications are induced by Forskolin, a cAMP analog. Signal transduction via the cAMP pathway is known to be ubiquitous and represents a major line of communication between many organisms and their environment. We believe that CCDC6 could be an important player in the dynamics of cAMP signaling by fine regulating CREB1 transcriptional activity in normal and transformed thyroid cells.
Sigma-Aldrich
Anti-CCDC6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
Sigma-Aldrich
Anti-CCDC6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1