Merck
  • EM523L, a nonpeptide motilin agonist, stimulates gastric emptying and pancreatic polypeptide secretion.
EM523L, a nonpeptide motilin agonist, stimulates gastric emptying and pancreatic polypeptide secretion.
Peptides (1996-01-01)
H Okano, A Inui, N Ueno, S Morimoto, A Ohmoto, M Miyamoto, N Aoyama, Y Nakajima, S Baba, M Kasuga
ABSTRACT
We investigated the efficacy and the mechanism of action of EM523L, a nonpeptide motilin agonist (motilide), on the stimulation of gastric emptying and on the release of gut peptides after ingestion of a solid meat in normal controls (n = 8) and in diabetic patients (n = 8) with signs of neuropathy. A dose of 2 mg EM523L was administered IV over 15 min just after ingestion of a solid meal (200 kcal Gastric emptying was measured by a radionuclide technique. EM523L accelerated gastric emptying and markedly augmented postprandial pancreatic polypeptide (PP) response in both normal control and diabetic patients. This may suggest the mediation of the Vagal-cholinergic pathway to accelerate gastric emptying. The present study offers a promising therapeutic potential of the motilide in gastrointestinal motility disorders like those observed in diabetics mellitus.
Product Description
Supelco
Erythromycin A enol ether, analytical standard