Merck
  • Effect of polyamidoamine dendrimer G3 and G4 on skin permeation of 8-methoxypsoralene--in vivo study.
Effect of polyamidoamine dendrimer G3 and G4 on skin permeation of 8-methoxypsoralene--in vivo study.
International journal of pharmaceutics (2012-02-09)
Katarzyna Borowska, Stanisław Wołowiec, Andrzej Rubaj, Kazimierz Głowniak, Elwira Sieniawska, Sebastian Radej
ABSTRACT
In the present study we have assessed the ability of (PAMAM) dendrimers G3 and G4 to facilitate transdermal delivery of 8-methoxypsoralen (8-MOP) in vivo. In vitro study using Franz diffusion cell revealed an enhanced transdermal flux for 8-MOP in complex with G3 and G4 dendrimer in relation to standard 8-MOP solution. In present study in vivo skin permeation potential of 8-MOP complex with G3 and G4 PAMAM dendrimer was assessed using confocal laser scanning microscopy (CLSM), which revealed an enhanced permeation of the 8-MOP to the deeper layers of the skin and significantly higher concentration in comparison with standard 8-MOP solution. Skin tissue 8-MOP concentration, evaluated by HPLC indicates that G3 and G4 PAMAM application significantly increase 8-MOP skin deposition in comparison with standard 8-MOP solutions after 1 and 2h. G4 appeared to be a more effective 8-MOP penetration enhancer than G3 PAMAM. Our results suggest the feasibility of G3 and G4 PAMAM dendrimers for transdermal delivery of 8-MOP resulting in better skin permeation and higher concentration of 8-MOP in epidermis and dermis of the drug that could help to improve effectiveness and safety of PUVA therapy.
Product Description
Supelco
8-Methoxypsoralen, analytical standard
Supelco
Xanthotoxin, analytical standard