Merck
  • Astrocyte Activation is A Potential Mechanism Underlying Depressed Mood and Apathy in People with HIV.
Astrocyte Activation is A Potential Mechanism Underlying Depressed Mood and Apathy in People with HIV.
Journal of neurology and psychology (2022-01-01)
Ronald J Ellis, Yan Fan, David Grelotti, Bin Tang, Scott Letendre, Johnny J He
ABSTRACT
Astrocytes become activated with certain infections, and this might alter the brain to trigger or worsen depressed mood. Indeed, astrocytes are chronically activated in people with HIV infection (PWH), who are much more frequently depressed than people without HIV (PWoH). A particularly disabling component of depression in PWH is apathy, a loss of interest, motivation, emotion, and goal-directed behavior. We tested the hypothesis that depression and apathy in PWH would be associated with higher levels of a biomarker of astrocyte activation, glial fibrillary acidic protein (GFAP), in cerebrospinal fluid (CSF). We evaluated PWH in a prospective observational study using the Beck Depression Inventory-II (BDI-II) and additional standardized assessments, including lumbar puncture. We measured GFAP in CSF with a customized direct sandwich ELISA method. Data were analyzed using ANOVA and multivariable regression. Participants were 212 PWH, mean (SD) age 40.9±9.14 years, median (IQR) nadir and current CD4 199 (57, 326) and 411 (259, 579), 65.1% on ART, 67.3% virally suppressed. Higher CSF GFAP correlated with worse total BDI-II total scores (Pearson correlation r=0.158, p-value=0.0211), and with worse apathy scores (r=0.205, p=0.0027). The correlation between apathy/depression and GFAP was not in fluenced by other factors such as age or HIV suppression status. Astrocyte activation, reflected in higher levels of CSF GFAP, was associated with worse depression and apathy in PWH. Interventions to reduce astrocyte activation -- for example, using a peptide-1 receptor (GLP-1R) agonist -- might be studied to evaluate their impact on disabling depression in PWH.
Sigma-Aldrich
Glial Fibrillary Acidic Protein, Human Brain