Merck
  • Multi-Stimuli-Responsive DOX Released from Magnetosome for Tumor Synergistic Theranostics.
Multi-Stimuli-Responsive DOX Released from Magnetosome for Tumor Synergistic Theranostics.
International journal of nanomedicine (2020-11-13)
Ming-Fong Tsai, Yu-Lun Lo, Yuan-Chun Huang, Chun-Chieh Yu, Yi-Ting Wu, Chia-Hao Su, Li-Fang Wang
ABSTRACT
To improve responses to tumor microenvironments for achieving a better therapeutic outcome in combination cancer therapy, poly(ε-caprolactone)-SS-poly(methacrylic acid) diblock copolymer (PCL-SS-PMAA) with a disulfide linkage between the hydrophobic and hydrophilic junctions was synthesized. Repeating units of PCL and PMAA in PCL-SS-PMAA were controlled and formulated into polymersomes (PSPps). Truncated octahedral Fe3O4 nanoparticles (IONPs) were synthesized and encapsulated to produce IONPs-PSPps NPs and doxorubicin (DOX) was further loaded to produce IONPs-PSPps@DOX NPs for theranostic applications. IONPs-PSPps NPs remained a superparamagnetic property with a saturation magnetization value of 85 emu⋅gFe3O4 -1 and a relaxivity value of 180 mM-1⋅s-1. Upon exposure to an alternating magnetic field (AMF), IONPs-PSPps NPs increased temperature from 25°C to 54°C within 15 min. Among test groups, the cell apoptosis was greatest in the group exposed to IONPs-PSPps@DOX NPs with AMF and magnet assistance. In vivo T2-weighted magnetic resonance images of A549 tumor-bearing mice also showed highest contrast and greatest tumor suppression in the tumor with AMF and magnet assistance. IONPs-PSPps@DOX NPs are a potential theranostic agent having multifaceted applications involving magnetic targeting, MRI diagnosis, hyperthermia and chemotherapy.
Sigma-Aldrich
1,1,4,7,10,10-Hexamethyltriethylenetetramine, 97%
Sigma-Aldrich
α-Bromoisobutyryl bromide, 98%
Sigma-Aldrich
tert-Butyl methacrylate, 98%, contains 200 ppm monomethyl ether hydroquinone as inhibitor