Merck
  • Pro-apoptotic activity of novel synthetic isoxazole derivatives exhibiting inhibitory activity against tumor cell growth in vitro.
Pro-apoptotic activity of novel synthetic isoxazole derivatives exhibiting inhibitory activity against tumor cell growth in vitro.
Oncology letters (2020-09-17)
Ilaria Lampronti, Daniele Simoni, Riccardo Rondanin, Riccardo Baruchello, Chiara Scapoli, Alessia Finotti, Monica Borgatti, Chiara Tupini, Roberto Gambari
ABSTRACT
In order to develop potential anticancer agents stimulating apoptosis, novel 3,4-isoxazolediamide and 4,5,6,7-tetrahydro-isoxazolo-[4,5-c]-pyridine derivatives have been synthetized. The original structures of geldanamycin and radicicol, which are known natural heat shock protein (HSP) inhibitors, were deeply modified because both of them exhibit several drawbacks, such as poor solubility, hepatotoxicity, intrinsic chemical instability or deprivation of the in vivo activity. This novel class of synthetic compounds containing the isoxazole nucleus exhibited potent and selective inhibition of HSP90 in previous studies. Biological assays (focusing on in vitro antiproliferative effects and pro-apoptotic activity) in human erythroleukemic K562 cells (as a model system referring to tumor cells grown in suspension), glioblastoma U251-MG and glioblastoma temozolomide (TMZ)-resistant T98G cell lines (two model systems referring to tumor cells grown attached to the flask), were performed. Almost all isoxazole derivatives demonstrated significant antiproliferative and pro-apoptotic activities, showing induction of both early and late apoptosis of K562 cells. Different effects were observed on the glioma U251-MG and T98G cells, depending on the structure of the analogues. Antiproliferative and pro-apoptotic activities in K562 cells were associated with the activation of the erythroid differentiation program. The present study demonstrated that 3,4-isoxazolediamide and 4,5,6,7-tetrahydro-isoxazolo-[4,5-c]-pyridine derivatives should be considered for in vivo studies focusing on the development of anticancer drugs acting, at least partially, via activation of apoptosis.
Sigma-Aldrich
Isoxazole, 99%