Merck
  • Korean Red Ginseng Extract Increases Apoptosis by Activation of the Noxa Pathway in Colorectal Cancer.
Korean Red Ginseng Extract Increases Apoptosis by Activation of the Noxa Pathway in Colorectal Cancer.
Nutrients (2019-09-01)
Yoon A Jeong, Bo Ram Kim, Dae Young Kim, Soyeon Jeong, Yoo Jin Na, Jung Lim Kim, Hye Kyeong Yun, Bu Gyeom Kim, Seong Hye Park, Min Jee Jo, Sun Il Lee, Byung-Cheol Han, Dae-Hee Lee, Sang Cheul Oh
ABSTRACT
Although the anticancer activity of Korean Red Ginseng (KRG) has been known in various cancers, the mechanism of KRG-induced apoptosis is unknown in colorectal cancer (CRC). In our study, we examined whether KRG induces apoptosis in CRC cells. In the cell viability assay, the concentration of the appropriate KRG extracts was fixed at 2.5 mg/mL in numerous CRC cells. This fixed concentration was in other experiments, and it was confirmed that the KRG extracts induce apoptosis in CRC cells. We found that KRG induced Noxa activation and apoptosis and increased endoplasmic reticulum stress via reactive oxygen species production. This indicated that KRG efficiently enhanced cell death in CRC cells. Our results show that KRG can be used as a possible anticancer drug for patients with CRC.
Sigma-Aldrich
MISSION® esiRNA, targeting human DDIT3
Sigma-Aldrich
MISSION® esiRNA, targeting human PMAIP1
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)