Merck
  • Design, Synthesis and Evaluation of N-pyrazinylbenzamides as Potential Antimycobacterial Agents.
Design, Synthesis and Evaluation of N-pyrazinylbenzamides as Potential Antimycobacterial Agents.
Molecules (Basel, Switzerland) (2018-09-21)
Jan Zitko, Alžběta Mindlová, Ondřej Valášek, Ondřej Jand'ourek, Pavla Paterová, Jiří Janoušek, Klára Konečná, Martin Doležal
ABSTRACT
Three series of N-(pyrazin-2-yl)benzamides were designed as retro-amide analogues of previously published N-phenylpyrazine-2-carboxamides with in vitro antimycobacterial activity. The synthesized retro-amides were evaluated for in vitro growth inhibiting activity against Mycobacterium tuberculosis H37Rv (Mtb), three non-tuberculous mycobacterial strains (M. avium, M. kansasii, M. smegmatis) and selected bacterial and fungal strains of clinical importance. Regarding activity against Mtb, most N-pyrazinylbenzamides (retro-amides) possessed lower or no activity compared to the corresponding N-phenylpyrazine-2-carboxamides with the same substitution pattern. However, the active retro-amides tended to have lower HepG2 cytotoxicity and better selectivity. Derivatives with 5-chloro substitution on the pyrazine ring were generally more active compared to their 6-cloro positional isomers or non-chlorinated analogues. The best antimycobacterial activity against Mtb was found in N-(5-chloropyrazin-2-yl)benzamides with short alkyl (2h: R² = Me; 2i: R² = Et) in position 4 of the benzene ring (MIC = 6.25 and 3.13 µg/mL, respectively, with SI > 10). N-(5-Chloropyrazin-2-ylbenzamides with hydroxy substitution (2b: R² = 2-OH; 2d: R² = 4-OH) on the benzene ring or their acetylated synthetic precursors possessed the broadest spectrum of activity, being active in all three groups of mycobacterial, bacterial and fungal strains. The substantial differences in in silico calculated properties (hydrogen-bond pattern analysis, molecular electrostatic potential, HOMO and LUMO) can justify the differences in biological activities between N-pyrazinylbenzamides and N-phenylpyrazine-2-carboxamides.
Sigma-Aldrich
Aminopyrazine, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2-amino-6-chloropyrazine, AldrichCPR
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2-amino-5-chloropyrazine, AldrichCPR