Merck
  • Dalbavancin: a new option for the treatment of gram-positive infections.
Dalbavancin: a new option for the treatment of gram-positive infections.
The Annals of pharmacotherapy (2006-03-02)
Shu-Wen Lin, Peggy L Carver, Daryl D DePestel
ABSTRACT
To review the pharmacology, microbiology, chemistry, in vitro susceptibility, pharmacokinetics, clinical efficacy, safety, tolerability, dosage, and administration of dalbavancin, a new semisynthetic lipoglycopeptide. A MEDLINE search, restricted to the English language, was conducted from 1966 through January 2006. Supplementary sources included program abstracts from the Interscience Conference on Antimicrobial Agents and Chemotherapy, American Society of Microbiology, and the Infectious Diseases Society of America from 2000 to 2005 and information available from the manufacturer's Web site. In vitro and preclinical studies, as well as Phase I, II, and III clinical trials, were evaluated to summarize the microbiology, pharmacology, clinical efficacy, and safety of dalbavancin. All published trials and abstracts citing dalbavancin were selected. Dalbavancin, a novel lipoglycopeptide, has a mechanism of action similar to that of other glycopeptides. It has in vitro activity against a variety of gram-positive organisms, but no activity against gram-negative or vancomycin-resistant enterococci that possess VanA gene. Due to its prolonged half-life (6-10 days), dalbavancin can be administered intravenously once weekly. In Phase II and III clinical trials, dalbavancin was effective and well tolerated for the treatment of skin and soft-tissue infections, catheter-related bloodstream infections, and skin and skin-structure infections. To date, adverse events are mild and limited; the most common include pyrexia, headache, nausea, oral candidiasis, diarrhea, and constipation. Dalbavancin appears to be a promising antimicrobial agent for the treatment of gram-positive infections. A new drug application was filed with the Food and Drug Administration (FDA) in December 2004. The FDA issued an approvable letter in 2005 for dalbavancin. If approved, dalbavancin is expected to be launched in the first quarter of 2006.
Sigma-Aldrich
Dalbavancin, ≥90% (HPLC)