
Giemsa stain solution

Synonym(s):
Giemsa stain, Azure mixture sicc. Giemsa stain
Empirical Formula (Hill Notation):
C14H14ClN3S
CAS Number:
Molecular Weight:
291.80
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Showing 1-1 of 1
Compare
Product Number
Description
Pricing