
Fluid Thioglycollate Medium (FTM)

Synonym(s):
Fluid Thioglycollate Medium (FTM)
Products
Product Category
Physical Form
Agency
Chemical Composition
Capacity (mL)
Quality Segment
Closure Type
Application
Package Size
Sterility
Sterilization Method
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing