Merck
Identification of a novel leukemic-specific splice variant of DNMT3B and its stability.
Medical oncology (Northwood, London, England) (2017-07-22)
Prachi Singh, Sarvagalla Sailu, Elango Palchamy, Mohane Selvaraj Coumar, Sudhakar Baluchamy
RESUMEN
DNA methyltransferases make use of alternative splicing mechanism to generate various splice variants, but their role(s) in modulating DNA methylation patterns in the cells is unclear. Notably, DNMT3B alone contains nearly 40 different splice variants. In this study, we have identified a novel splice variant of DNMT3B, which lacks exon 7 and 10 from leukemic cell lines which we termed as DNMT3B9. The exon 7 codes for the major part of PWWP domain, and exon 10 inclusion serves as a pluripotent marker. By quantitative RT-PCR using exon-exon junction-specific primers, we showed higher level of DNMT3B9 transcripts in several leukemic cell lines. However, DNMT3B9 expression was less in other tested cancer cell lines indicating that DNMT3B9 might serve as a leukemic-specific biomarker. Surprisingly, endogenous protein for DNMT3B9 was not detected in leukemic cells suggesting the unidentified RNA-related function(s) for DNMT3B9. In addition, we showed that DNMT3B9 protein lacks PWWP domain is less stable compared to other DNMT3B variants which contain PWWP domain through computational predictions and by cycloheximide half-life experiment. Taken together, we demonstrated the existence of novel leukemic-specific splice variant of DNMT3B and provide the evidence for the role of PWWP domain in the stability of DNMT3B. The findings reported here have relevance in epigenetic therapy, which is aimed to target the DNMT3B in cancer cells.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Monoclonal Anti-T7 tag antibody produced in mouse, purified immunoglobulin, buffered aqueous solution