Merck
Disruption of the C. elegans Intestinal Brush Border by the Fungal Lectin CCL2 Phenocopies Dietary Lectin Toxicity in Mammals.
PloS one (2015-06-10)
Katrin Stutz, Andres Kaech, Markus Aebi, Markus Künzler, Michael O Hengartner
RESUMEN
Lectins are non-immunoglobulin carbohydrate-binding proteins without enzymatic activity towards the bound carbohydrates. Many lectins of e.g. plants or fungi have been suggested to act as toxins to defend the host against predators and parasites. We have previously shown that the Coprinopsis cinerea lectin 2 (CCL2), which binds to α1,3-fucosylated N-glycan cores, is toxic to Caenorhabditis elegans and results in developmental delay and premature death. In this study, we investigated the underlying toxicity phenotype at the cellular level by electron and confocal microscopy. We found that CCL2 directly binds to the intestinal apical surface and leads to a highly damaged brush border with loss of microvilli, actin filament depolymerization, and invaginations of the intestinal apical plasma membrane through gaps in the terminal web. We excluded several possible toxicity mechanisms such as internalization and pore-formation, suggesting that CCL2 acts directly on intestinal apical plasma membrane or glycocalyx proteins. A genetic screen for C. elegans mutants resistant to CCL2 generated over a dozen new alleles in bre 1, ger 1, and fut 1, three genes required for the synthesis of the sugar moiety recognized by CCL2. CCL2-induced intestinal brush border defects in C. elegans are similar to the damage observed previously in rats after feeding the dietary lectins wheat germ agglutinin or concanavalin A. The evolutionary conserved reaction of the brush border between mammals and nematodes might allow C. elegans to be exploited as model organism for the study of dietary lectin-induced intestinal pathology in mammals.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
DIC, 99%
Sigma-Aldrich
Metanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Acetona, JIS special grade, ≥99.5%
Sigma-Aldrich
Metanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Metanol, anhydrous, 99.8%
Sigma-Aldrich
Acetona, natural, ≥97%
Sigma-Aldrich
Acetona, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetona, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Metanol, suitable for HPLC
Sigma-Aldrich
Metanol, SAJ special grade
Sigma-Aldrich
Acetona, ≥99.5%, for residue analysis
Sigma-Aldrich
Metanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Metanol, NMR reference standard
Sigma-Aldrich
Acetona, for residue analysis, ≥99.5%
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Diisopropylcarbodiimide solution, 1 M in THF
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Acetona, for chromatography, ≥99.8%
Sigma-Aldrich
Acetona, for residue analysis, JIS 5000
Sigma-Aldrich
Acetona, suitable for HPLC, ≥99.9%
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Metanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Acetona, suitable for HPLC
Sigma-Aldrich
Metanol, HPLC Plus, ≥99.9%, poly-coated bottles