Merck
Separation of aromatic hydrophobic sulfonates by micellar electrokinetic chromatography.
Journal of chromatography. A (2007-07-14)
Sille Ehala, Merike Vaher, Mihkel Kaljurand
RESUMEN
Two different buffer systems for the separation of 12 aromatic hydrophobic sulfonates by micellar electrokinetic chromatography (MEKC) were developed. The following buffer systems were used: aqueous phosphate buffers containing either cetyltrimethylammonium bromide (CTAB) or sodium dodecyl sulfate (SDS). Eleven aromatic sulfonates were simultaneously separated in less than 35 min employing 20 mM phosphate buffer, pH 7.0 containing 50 mM SDS and 10% of acetonitrile.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
2-Chloroethyl p-toluenesulfonate, 97%