Merck
  • Discovery of thiazolidine-2,4-dione/biphenylcarbonitrile hybrid as dual PPAR α/γ modulator with antidiabetic effect: in vitro, in silico and in vivo approaches.
Discovery of thiazolidine-2,4-dione/biphenylcarbonitrile hybrid as dual PPAR α/γ modulator with antidiabetic effect: in vitro, in silico and in vivo approaches.
Chemical biology & drug design (2013-01-08)
Sergio Hidalgo-Figueroa, Juan J Ramírez-Espinosa, Samuel Estrada-Soto, Julio C Almanza-Pérez, Rubén Román-Ramos, Francisco J Alarcón-Aguilar, Jesús V Hernández-Rosado, Hermenegilda Moreno-Díaz, Daniel Díaz-Coutiño, Gabriel Navarrete-Vázquez
RESUMEN
A small series of thiazolidine-2,4-dione and barbituric acid derivatives 1-4 was prepared using a short synthetic route, and all compounds were characterized by elemental analysis, mass spectrometry, and NMR ((1)H, (13)C) spectroscopy. Their in vitro relative expression of peroxisome proliferator-activated receptor α and peroxisome proliferator-activated receptor γ was evaluated. Compound 1 showed an increase in the mRNA expression of both peroxisome proliferator-activated receptor isoforms, as well as the GLUT-4 levels. The antidiabetic activity of compound 1 was determined at 50 mg/kg single dose using a non-insulin-dependent diabetes mellitus rat model. The results indicated a significant decrease in plasma glucose levels. Additionally, we performed a molecular docking of compound 1 into the ligand binding pocket of peroxisome proliferator-activated receptor α and peroxisome proliferator-activated receptor γ. In these binding models, compound 1 may bind into the active site of both isoforms showing important short contacts with the peroxisome proliferator-activated receptor γ residues: Tyr 473, His 449, Ser 289, His 323; and peroxisome proliferator-activated receptor α residues: Tyr 464, His 440, Ser 280 and Tyr 314.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Barbituric acid, ReagentPlus®, 99%
Supelco
Barbituric acid, for spectrophotometric det. of cyanide, ≥99.5%