Merck
Effect of maturity, processing, and storage on the furanocoumarin composition of grapefruit and grapefruit juice.
Journal of food science (2012-03-16)
Paul F Cancalon, Santiago M Barros, Carl Haun, Wilbur W Widmer
RESUMEN
Since the early 1990's, grapefruit juice has been implicated in drug interaction with various furanocoumarins (FCs) now associated with the effect. Although FCs are present in various fruits and vegetables, it is their presence in grapefruit that has attracted the most attention. Studies have shown that FCs in grapefruit juice can vary significantly and from multiple causes. Most of all, FCs are stress-induced molecules, their levels affected by many factors ranging from UV exposure to insect infestation. There are also varietal and seasonal factors. In this study, juice processing and storage parameters were investigated. Prolonged fruit storage prior to processing and most steps involved in juice processing had little influence on the levels of 6',7'-dihydroxybergamottin (DHB), paradisin C, or bergamottin. However, products that were hot filled or stored at room temperature had lower amounts of DHB and paradisin C and higher amounts of bergaptol compared to juices that were not hot filled and stored at refrigerated temperatures. Both DHB and paradisin C are potent CYP3A4 inhibitors, while bergaptol is a very weak inhibitor. Bergamottin amounts decreased to a lesser extent. Therefore, grapefruit juice products that were hot filled or have been stored at room temperature for an extended period of time will have a reduced drug interaction potential.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Supelco
Bergamottin, analytical standard