Merck
Peptide receptor radionuclide therapy (PRRT) for GEP-NETs.
Best practice & research. Clinical gastroenterology (2013-04-16)
Hendrik Bergsma, Esther I van Vliet, Jaap J M Teunissen, Boen L R Kam, Wouter W de Herder, Robin P Peeters, Eric P Krenning, Dik J Kwekkeboom
RESUMEN
Peptide receptor radionuclide therapy (PRRT) with radiolabelled somatostatin analogues plays an increasing role in the treatment of patients with inoperable or metastasised gatroenteropancreatic neuroendocrine tumours (GEP-NETs). (90)Y-DOTATOC and (177)Lu-DOTATATE are the most used radiopeptides for PRRT with comparable tumour response rates (about 15-35%). The side effects of this therapy are few and mild. However, amino acids should be used for kidney protection, especially during infusion of (90)Y-DOTATOC. Options to improve PRRT may include combinations of radioactive labelled somatostatin analogues and the use of radiosensitising drugs combined with PRRT. Other therapeutic applications of PRRT may include intra-arterial administration, neo-adjuvant treatment and additional PRRT cycles in patients with progressive disease, who have benefited from initial therapy. Considering the mild side-effects, PRRT may well become the first-line therapy in patients with metastasised or inoperable GEP-NETs if more widespread use of PRRT can be accomplished.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Octreotide, ≥98% (HPLC)