Merck
Therapeutic significance of estrogen receptor β agonists in gliomas.
Molecular cancer therapeutics (2012-03-24)
Gangadhara R Sareddy, Binoj C Nair, Vijay K Gonugunta, Quan-guang Zhang, Andrew Brenner, Darrell W Brann, Rajeshwar Rao Tekmal, Ratna K Vadlamudi
RESUMEN
Gliomas are the most common and devastating central nervous system neoplasms. A gender bias exists in their development: females are at lower risk than males, implicating estrogen-mediated protective effects. Estrogen functions are mediated by two estrogen receptor (ER) subtypes: ERα, which functions as tumor promoter, and ERβ, which functions as tumor suppressor. We examined the potential use of ERβ agonists as a novel therapeutic to curb the growth of gliomas. Western analysis of six glioma model cells showed detectable expression of ERβ with little or no ERα. Treatment of glioma cells with ERβ agonists resulted in significant decrease in proliferation. Immunohistochemical analysis of tumor tissues revealed that ERβ expression is downregulated in high-grade gliomas. We found that ERβ agonists promote both expression and tumor-suppressive functions of ERβ in glioma cells. Liquiritigenin, a plant-derived ERβ agonist significantly reduced in vivo tumor growth in a xenograft model. Compared with control mice, animals treated with liquiritigenin had greater than 50% reduction in tumor volume and size. Immunohistochemical analysis of tumors revealed a significant increase in the nuclear ERβ expression with a concomitant decrease in cell proliferation in the liquiritigenin-treated group. Our results suggest that ERβ signaling has a tumor-suppressive function in gliomas. Because ERβ agonists are currently in clinical trials and are well tolerated with fewer side effects, identification of an ERβ agonist as a therapeutic agent can be readily extended to clinical use with current chemotherapies, providing an additional tool for enhancing survival in glioma patients.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Liquiritigenin, ≥97.0% (HPLC)