Merck
  • Reduction of N-glycolylneuraminic acid in human induced pluripotent stem cells generated or cultured under feeder- and serum-free defined conditions.
Reduction of N-glycolylneuraminic acid in human induced pluripotent stem cells generated or cultured under feeder- and serum-free defined conditions.
PloS one (2010-12-03)
Yohei Hayashi, Techuan Chan, Masaki Warashina, Masakazu Fukuda, Takashi Ariizumi, Koji Okabayashi, Naoya Takayama, Makoto Otsu, Koji Eto, Miho Kusuda Furue, Tatsuo Michiue, Kiyoshi Ohnuma, Hiromitsu Nakauchi, Makoto Asashima
RESUMEN
The successful establishment of human induced pluripotent stem cells (hiPSCs) has increased the possible applications of stem cell research in biology and medicine. In particular, hiPSCs are a promising source of cells for regenerative medicine and pharmacology. However, one of the major obstacles to such uses for hiPSCs is the risk of contamination from undefined pathogens in conventional culture conditions that use serum replacement and mouse embryonic fibroblasts as feeder cells. Here we report a simple method for generating or culturing hiPSCs under feeder- and serum-free defined culture conditions that we developed previously for human embryonic stem cells. The defined culture condition comprises a basal medium with a minimal number of defined components including five highly purified proteins and fibronectin as a substrate. First, hiPSCs, which were generated using Yamanaka's four factors and conventional undefined culture conditions, adapted to the defined culture conditions. These adapted cells retained the property of self renewal as evaluated morphologically, the expression of self-renewal marker proteins, standard growth rates, and pluripotency as evaluated by differentiation into derivatives of all three primary germ layers in vitro and in vivo (teratoma formation in immunodeficient mice). Moreover, levels of nonhuman N-glycolylneuraminic acid (Neu5Gc), which is a xenoantigenic indicator of pathogen contamination in human iPS cell cultures, were markedly decreased in hiPSCs cultured under the defined conditions. Second, we successfully generated hiPSCs using adult dermal fibroblast under the defined culture conditions from the reprogramming step. For a long therm culture, the generated cells also had the property of self renewal and pluripotency, they carried a normal karyotype, and they were Neu5Gc negative. This study suggested that generation or adaption culturing under defined culture conditions can eliminate the risk posed by undefined pathogens. This success in generating hiPSCs using adult fibroblast would be beneficial for clinical application.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Apo-transferrina human, powder, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
N-Glycolylneuraminic acid, ≥95% (HPLC), semisynthetic