Merck
The Family Secrets of Avian Egg-Specific Ovalbumin and Its Related Proteins Y and X.
Biology of reproduction (2015-07-15)
Mylene Da Silva, Stéphane Beauclercq, Grégoire Harichaux, Valérie Labas, Nicolas Guyot, Joel Gautron, Yves Nys, Sophie Rehault-Godbert
RESUMEN
The ovalbumin gene family in Gallus gallus is composed of three homologous genes located within a 46 kb locus on chromosome 2: ovalbumin, ovalbumin-related protein Y (OVAY), and ovalbumin-related protein X (OVAX) genes. The expression of these genes in hen oviduct is under estrogen control, but their relative hormonal responsiveness and subsequent protein concentration in egg, is distinctive. Interestingly, all three proteins lack the classical signal peptide for secretion. Ovalbumin, OVAX, and OVAY belong to the serine protease inhibitor (serpin) family whose members share a common tertiary structure. Ovalbumin and OVAX are one of the few members of this family that do not express any protease inhibition activity whereas OVAY has been predicted to be inhibitory, by comparison with the consensus sequence for inhibitory serpins. In contrast to ovalbumin and OVAY, OVAX interacts with heparin, a negatively charged glycosaminoglycan, via a positively charged domain exposed at the surface of the molecule. Ovalbumin is the major egg white protein and might be a source of amino acids for the developing embryo. The physiological function of OVAY is not known, but recent data have revealed a possible role of this protein in early embryonic development. Considering the antibacterial activities of OVAX, this protein might play a role in egg defense. This review sheds light on the expression, biochemistry, and structural specificities of these three highly similar paralogs. It gives new clues in favor of diverging functions, which are likely to have arisen by duplication events from a common ancestral gene.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Albumin hydrolysate