Merck
Exploiting gelatin nanocarriers in the pulmonary delivery of methotrexate for lung cancer therapy.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2019-03-25)
Hend Abdelrady, Rania M Hathout, Rihab Osman, Imran Saleem, Nahed D Mortada
RESUMEN
Gelatin has many merits that encourage its use in the pulmonary delivery of anticancer drugs. It is a biodegradable denatured protein which possesses several functional groups that could be modified. Additionally, it has balanced hydrophilic and hydrophobic characters, which facilitate the loading of chemotherapeutic agents. Accordingly, the purpose of the current work was to exploit this valuable biomaterial in the efficient pulmonary delivery of methotrexate in case of lung cancer. Gelatin nanoparticles were prepared via a desolvation method and the fabrication process was optimized using Box Behnken design of experiment. A comparative study on uptake of gelatin nanoparticles by lung adenocarcinoma cells and macrophages was implemented using flow cytometry. Investigation of the effect of different methotrexate loading techniques: encapsulation, post loading and chemical conjugation on the nanoparticles characteristics and cellular cytotoxicity was performed. Nano-in-microparticles were prepared by co-spray drying optimized nanoparticles with leucine. Results showed that Box Behnken design was able to optimize preparation parameters to yield uniform nanoparticles with suitable particle size for cancer cells uptake. The prepared nanoparticles demonstrated a preferential uptake by lung cancer cells. Additionally, methotrexate loaded nanoparticles demonstrated up to four fold significant reduction in methotrexate IC50. The spray dried gelatin nano-in microparticles demonstrated good aerosolization properties enabling lung deposition in the respirable airways. Thus, providing a promising platform for lung cancer therapy.