Merck
  • Design, synthesis and aromatase inhibitory activities of novel indole-imidazole derivatives.
Design, synthesis and aromatase inhibitory activities of novel indole-imidazole derivatives.
Bioorganic & medicinal chemistry letters (2013-02-14)
Rui Wang, Hong-Fan Shi, Jing-Feng Zhao, Yan-Ping He, Hong-Bin Zhang, Jian-Ping Liu
ABSTRACT
A series of novel indole-imidazole derivatives have been prepared and evaluated in vitro on the aromatase inhibitory activities. The results suggested that proton or a small electron-withdrawing group at para-position of the phenyl ring would enhance the inhibitory activities and any bulky group should be avoided in order to keep a relative small volume for this kind of molecules.
Product Description
Imidazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Indole, ≥99%
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Imidazole hydrochloride
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Indole, ≥99%, FG
Sigma-Aldrich
Imidazole sodium derivative, technical grade
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O