Merck
Biocomposite films based on alginate and organically modified clay.
International journal of biological macromolecules (2012-01-25)
Filiz Tezcan, Ebru Günister, Gönül Ozen, F Bedia Erim
ABSTRACT
Sodium alginate/sodium montmorillonite hybrid films were prepared by casting from the suspension of sodium alginate and different clay samples. Clay samples had been modified with a cationic surfactant, a cationic polymer, and a small polar molecule, respectively. Benzethonium chloride, polyethyleneimine and urea were used as clay modifiers. The composite films begin to disintegrate at a higher temperature and with less weight loss than the pure alginate films. This suggests an enhancement of the film thermal stability due to the modification of the alginate with clay samples.
Sigma-Aldrich
Hyamine® 1622 solution, 4 mM in H2O
Sigma-Aldrich
Benzethonium chloride, ≥97% (titration), ≥98% (HPLC)
Sigma-Aldrich
Benzethonium chloride, BioUltra, ≥99.0% (AT)