Merck
  • Unobstructed Ultrathin Gas Transport Channels in Composite Membranes by Interfacial Self-Assembly.
Unobstructed Ultrathin Gas Transport Channels in Composite Membranes by Interfacial Self-Assembly.
Advanced materials (Deerfield Beach, Fla.) (2020-04-25)
Bo Wang, Zhihua Qiao, Jiayou Xu, Jixiao Wang, Xinlei Liu, Song Zhao, Zhi Wang, Michael D Guiver
ABSTRACT
Ultrathin unobstructed gas transport channels through the membrane selective layer are constructed in mixed matrix membranes (MMMs) by using gravity-induced interface self-assembly of poly(vinylamine) and polymer-modified MIL-101(Cr). For CO2 /N2 (15/85 by volume) mixed gas, the MMMs achieve a high CO2 permeance of 823 gas permeation units and CO2 /N2 selectivity of 242 at 0.5 MPa. Based on economic analyses, a two-stage membrane process can achieve gas separation and economic targets.
Sigma-Aldrich
N-Vinylformamide, 98%