Merck
  • Breakpoints for susceptibility testing should not divide wild-type distributions of important target species.
Breakpoints for susceptibility testing should not divide wild-type distributions of important target species.
Antimicrobial agents and chemotherapy (2009-02-04)
Maiken Cavling Arendrup, Gunnar Kahlmeter, Juan Luis Rodriguez-Tudela, J Peter Donnelly
ABSTRACT
The fluconazole MIC distributions for Candida glabrata from testing 34 different clinical isolates and performing 51 tests on a single isolate mirrored each other. Since what is perceived as biological variation in isolates without resistance mechanisms is mainly methodological variation, breakpoints which divide this distribution not only lack a sound biological basis but also result in poor reproducibility of susceptibility characterization. This makes 2, 4, 8, and possibly 16 microg/ml unsuitable breakpoints for C. glabrata and fluconazole.
Sigma-Aldrich
Fluconazole, ≥98% (HPLC), powder