Merck
  • SKI3301, a purified herbal extract from Sophora tonkinensis, inhibited airway inflammation and bronchospasm in allergic asthma animal models in vivo.
SKI3301, a purified herbal extract from Sophora tonkinensis, inhibited airway inflammation and bronchospasm in allergic asthma animal models in vivo.
Journal of ethnopharmacology (2017-05-17)
Hunseung Yoo, Minseok Kang, Sungsoo Pyo, Hee-Sung Chae, Keun Ho Ryu, Jinwoong Kim, Young-Won Chin
ABSTRACT
Sophora tonkinensis (Leguminosae, ST) is a traditional herbal plant in Korea and China. Its roots and rhizomes have been used to dissipate heat, to clear toxic material and to treat acute pharyngolaryngeal infections and sore throats. In this study, we tried to investigate the anti-inflammatory and anti-asthmatic effects of a purified extract (SKI3301) from Sophora tonkinensis using in vitro enzyme assay models and ovalbumin (OVA)-induced asthma animal models. The effect of SKI3301 on pro-inflammatory enzymes such as 5-lipoxygenase, phosphodiesterase 3 & 4, and thromboxane synthase was assayed in vitro. BALB/c mice were sensitized with OVA/Alum ip injection and nebulized with OVA to induce airway inflammation. Bronchoalveolar lavage (BAL) fluid was collected and analyzed for leukocytes infiltration and IL-5 production along with lung histopathology. Guinea pigs passively sensitized with anti-OVA antiserum were used to investigate the effect of SKI3301 on bronchospasm in vitro and in vivo. SKI3301 potently inhibited the activities of 5-lipoxygenase, phosphodiesterase 3 & 4, and thromboxane synthase. Orally administered SKI3301 attenuated the total leukocytes and eosinophil infiltration and IL-5 level in BAL fluids. Histopathological changes associated with lung inflammation were also reduced by SKI3301. SKI3301 inhibited OVA-induced contraction of isolated trachea from sensitized guinea pigs. SKI3301 also protected OVA-induced bronchoconstriction in the sensitized guinea pigs. Maackiain, one of 3 major components of SKI3301, was effective in inhibiting 5-lipoxygenase and OVA-induced airway inflammation. In this study, SKI3301 potently inhibited pro-inflammatory enzymes and attenuated OVA-induced bronchospasm in animal model of allergic asthma. These results suggest that SKI3301 may have therapeutic potential for allergic asthma.
Sigma-Aldrich
Trifolirhizin, ≥95% (LC/MS-ELSD)
Sigma-Aldrich
Snake venom from Crotalus atrox (Western Diamondback Rattlesnake)