Merck
  • Dietary Protein to Carbohydrate Ratio and Caloric Restriction: Comparing Metabolic Outcomes in Mice.
Dietary Protein to Carbohydrate Ratio and Caloric Restriction: Comparing Metabolic Outcomes in Mice.
Cell reports (2015-06-02)
Samantha M Solon-Biet, Sarah J Mitchell, Sean C P Coogan, Victoria C Cogger, Rahul Gokarn, Aisling C McMahon, David Raubenheimer, Rafael de Cabo, Stephen J Simpson, David G Le Couteur
ABSTRACT
Both caloric restriction (CR) and low-protein, high-carbohydrate (LPHC) ad-libitum-fed diets increase lifespan and improve metabolic parameters such as insulin, glucose, and blood lipids. Severe CR, however, is unsustainable for most people; therefore, it is important to determine whether manipulating macronutrient ratios in ad-libitum-fed conditions can generate similar health outcomes. We present the results of a short-term (8 week) dietary manipulation on metabolic outcomes in mice. We compared three diets varying in protein to carbohydrate ratio under both CR and ad libitum conditions. Ad libitum LPHC diets delivered similar benefits to CR in terms of levels of insulin, glucose, lipids, and HOMA, despite increased energy intake. CR on LPHC diets did not provide additional benefits relative to ad libitum LPHC. We show that LPHC diets under ad-libitum-fed conditions generate the metabolic benefits of CR without a 40% reduction in total caloric intake.
Sigma-Aldrich
Monoclonal Anti-Insulin antibody produced in mouse, clone K36AC10, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Glucagon antibody produced in mouse, clone K79bB10, ascites fluid
Sigma-Aldrich
Anti-Mouse IgG1 (γ-chain specific)-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder