Merck
  • [Effects of oxygen and nikethamide on central drive, ventilation and blood gases of patients with obstructive lung disease in acute exacerbation of respiratory failure].
[Effects of oxygen and nikethamide on central drive, ventilation and blood gases of patients with obstructive lung disease in acute exacerbation of respiratory failure].
Zhonghua jie he he hu xi za zhi = Zhonghua jiehe he huxi zazhi = Chinese journal of tuberculosis and respiratory diseases (1993-12-01)
X H Ren, D J Ding, E Z Chen
ABSTRACT
Twelve subjects with COPD in acute exacerbation of respiratory failure were studied. The experiment of each subject was divided into three steps: room air breathing, 35% O2 inhalation for one hour, and then intravenous drip of nikethamide (1.875g) for two hours with 35% oxygen inhalation at the same time. At the end of each step, mouth occlusion pressure (P0.1), VT, VE, VA, VCO2, VD and PaO2, PaCO2 were measured respectively. The results showed that, when breathing air, all the patients presented significant higher P0.1 than normal subjects, indicated higher central drive. After oxygen inhalation, P0.1 decreased markedly, but still higher than normal. No correlation was found between delta P0.1 and delta PaO2. VE declined with the drop of P0.1, but this was due to a decrease of respiratory frequency, while VA remained unchanged (P > 0.05). The increase of PaCO2 was unremarkable. Neither correlation was found between delta VA and delta P0.1, nor between delta VA and delta PaCO2. However, a close correlation existed between delta VCO2/VCO2 and delta VE/VE. The result of our study is not consistent with the postulation, the removal of the hypoxic stimulate after oxygen administration results in a decrease of ventilation and CO2 retention. After nikethamide administration, P0.1 increased as well as VE while VA and PaCO2 remained unchanged. The increase of VE was caused by the increase of respiratory rate. Furthermore, PaO2 decreased in some patients. All of the changes demonstrated that nothing is worthwhile with the treatment of nikethamide, but a side effect from increasing work of breathing and consumption of oxygen.(ABSTRACT TRUNCATED AT 250 WORDS)
Sigma-Aldrich
N,N-Diethylnicotinamide, 99%