Merck
  • Regioselective catalytic hydroboration of propargylic species using Cu(I)-NHC complexes.
Regioselective catalytic hydroboration of propargylic species using Cu(I)-NHC complexes.
Organic letters (2012-09-06)
Jin Kyoon Park, Brian A Ondrusek, D Tyler McQuade
ABSTRACT
The catalytic regioselective hydroboration of propargylic alcohols and ethers was investigated using NHC-CuCl. We observe that different NHC-CuCl complexes catalyze hydroborations of propargylic substrates with opposite regioselectivity. A 6-NHC-CuCl complex provides α-selectivity whereas β-selectivity is achieved using a 5-NHC-CuCl complex. The reaction tolerates a wide range of functional groups.
Sigma-Aldrich
Propargyl alcohol, 99%