Merck
  • A porous metal-organic replica of α-PbO2 for capture of nerve agent surrogate.
A porous metal-organic replica of α-PbO2 for capture of nerve agent surrogate.
Journal of the American Chemical Society (2010-12-09)
Ruqiang Zou, Ruiqin Zhong, Songbai Han, Hongwu Xu, Anthony K Burrell, Neil Henson, Jonathan L Cape, Donald D Hickmott, Tatiana V Timofeeva, Toti E Larson, Yusheng Zhao
ABSTRACT
A novel metal-organic replica of α-PbO(2) exhibits high capacity for capture of nerve agent surrogate.
Sigma-Aldrich
Lead(II) oxide, SAJ first grade, ≥98.0%
Sigma-Aldrich
Lead(II) oxide, 99.999% trace metals basis
Sigma-Aldrich
Lead(II) oxide, powder, <10 μm, ReagentPlus®, ≥99.9% trace metals basis
Sigma-Aldrich
Lead(II) oxide, ACS reagent, ≥99.0%
Sigma-Aldrich
Lead(II) oxide, puriss. p.a., ≥99.0% (KT), yellow