Merck
  • pH-controlled change of the metal coordination in a dicopper(II) complex of the ligand H-BPMP: crystal structures, magnetic properties, and catecholase activity.
pH-controlled change of the metal coordination in a dicopper(II) complex of the ligand H-BPMP: crystal structures, magnetic properties, and catecholase activity.
Inorganic chemistry (2001-02-24)
S Torelli, C Belle, I Gautier-Luneau, J L Pierre, E Saint-Aman, J M Latour, L Le Pape, D Luneau
ABSTRACT
The dinucleating ligand 2,6-bis[(bis(2-pyridylmethyl)amino)methyl]-4-methylphenol (H-BPMP) has been used to synthesize the three dinuclear Cu(II) complexes [Cu2(BPMP)(OH)][ClO4](2).0.5C4H8O (1), [Cu2(BPMP)(H2O)2](ClO4)(3).4H2O (2), and [Cu2(H-BPMP)][(ClO4)4].2CH3CN (3). X-ray diffraction studies reveal that 1 is a mu-hydroxo, mu-phenoxo complex, 2 a diaqua, mu-phenoxo complex, and 3 a binuclear complex with Cu-Cu distances of 2.96, 4.32, and 6.92 A, respectively. Magnetization measurements reveal that 1 is moderately antiferromagnetically coupled while 2 and 3 are essentially uncoupled. The electronic spectra in acetonitrile or in water solutions give results in accordance with the solid-state structures. 1 is EPR-silent, in agreement with the antiferromagnetic coupling between the two copper atoms. The X-band spectrum of powdered 2 is consistent with a tetragonally elongated square pyramid geometry around the Cu(II) ions, in accordance with the solid-state structure, while the spectrum in frozen solution suggests a change in the coordination geometry. The EPR spectra of 3 corroborate the solid-state and UV-visible studies. The 1H NMR spectra also lead to observations in accordance with the conclusions from other spectroscopies. The electrochemical behavior of 1 and 2 in acetonitrile or in water solutions shows that the first reduction (Cu(II)Cu(II)-Cu(II)Cu(I) redox couple) is reversible and the second (formation of Cu(I)Cu(I) irreversible. In water, 1 and 2 are reversibly interconverted upon acid/base titration (pK 4.95). In basic medium a new species, 4, is reversibly formed (pK 12.0), identified as the bishydroxo complex. Only 1 exhibits catecholase activity (oxidation of 3,5-di-tert-butylcatechol to the corresponding quinone, vmax = 1.1 x 10(-6) M-1 s-1 and KM = 1.49 mM). The results indicate that the pH dependence of the catalytic abilities of the complexes is related to changes in the coordination sphere of the metal centers.
Sigma-Aldrich
3,5-Di-tert-butylcatechol, 98%