Merck
  • Receptor-interacting protein shuttles between cell death and survival signaling pathways.
Receptor-interacting protein shuttles between cell death and survival signaling pathways.
Molecular biology of the cell (2009-12-04)
Pachiyappan Kamarajan, Julius Bunek, Yong Lin, Gabriel Nunez, Yvonne L Kapila
ABSTRACT
Cross-talk between apoptosis and survival signaling pathways is crucial for regulating tissue processes and mitigating disease. We report that anoikis-apoptosis triggered by loss of extracellular matrix contacts-activates a CD95/Fas-mediated signaling pathway regulated by receptor-interacting protein (RIP), a kinase that shuttles between CD95/Fas-mediated cell death and integrin/focal adhesion kinase (FAK)-mediated survival pathways. RIP's death domain was critical for RIP and Fas association to mediate anoikis. Fas or RIP attenuation reduced this association and suppressed anoikis, whereas their overexpression had the reverse effect. Overexpressing FAK restored RIP and FAK association and inhibited anoikis. Thus, RIP shuttles between CD95/Fas death and FAK survival signaling to mediate anoikis.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution