Merck
IL-35 is a Protective Immunomodulator in Brain Ischemic Injury in Mice.
Neurochemical research (2018-06-20)
Chen Xu, Hao Zhu, Rong Shen, Qian Feng, Hua Zhou, Zhong Zhao
ABSTRACT
IL-35 has been identified as a novel anti-inflammatory cytokine that belongs to the IL-12 cytokine family and has been verified to play a protective role in autoimmune diseases. In this study, we investigated the protective effects of IL-35 on cerebral ischemia/reperfusion (I/R) injury in a middle cerebral artery occlusion mouse model. We determined that the expression of IL-35 was initially decreased and subsequently increased in I/R injury. Moreover, IL-35 (i.c.v.) pre- and posttreatment significantly reduced the infarct volume and improved neurological deficits after 45 min of ischemia and 24 h of reperfusion. Importantly, IL-35 treatment improved neurological function recovery, particularly in balance ability, at 14 days after treatment. Finally, our results showed that IL-35 treatment reduced the expression of IL-6 and IL-1β, which are confirmed proinflammatory cytokines, thus indicating that these cytokines have both been linked to the anti-inflammatory mechanisms of IL-35. Therefore, IL-35 may be a key immune mediator in brain ischemic injury and appears to have promising potential for clinical trials.
Sigma-Aldrich
IL-35 (mouse): FC (human), recombinant, expressed in CHO cells