
Stavudine for system suitability

Synonym(s):
2′,3′-Didehydro-3′-deoxythymidine, 1-(2,3-Dideoxy-β-D-glycero-pent-2-enofuranosyl)thymine, 1-[(2R,5S)-5-(Hydroxymethyl)-2,5-dihydrofuran-2-yl]-5-methylpyrimidine-2,4-(1H,3H)-dione, 2′,3′-Anhydrothymidine, Stavudine, d4T
Empirical Formula (Hill Notation):
C10H12N2O4
CAS Number:
Molecular Weight:
224.21
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing