
(R)-(+)-SKF-38393 hydrochloride

Empirical Formula (Hill Notation):
C16H17NO2 · HCl
CAS Number:
Molecular Weight:
291.77
Products
Product Category
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing