![NovaSeptum® GO Preloaded System with High Purity Bag, 9-port sterile; β-irradiated](/deepweb/assets/sigmaaldrich/product/images/135/323/5f954531-baa6-45f4-b3b2-5696e14f5db3/640/5f954531-baa6-45f4-b3b2-5696e14f5db3.jpg)
NovaSeptum® GO Preloaded System with High Purity Bag, 9-port
![NovaSeptum® GO Preloaded System with High Purity Bag, 9-port sterile; β-irradiated](/deepweb/assets/sigmaaldrich/product/images/135/323/5f954531-baa6-45f4-b3b2-5696e14f5db3/640/5f954531-baa6-45f4-b3b2-5696e14f5db3.jpg)
Synonym(s):
NovaSeptum® GO High Purity Bag
Products
Product Category
Brand
Product Line
Agency
Quality Segment
Application
Sterility
Sterilization Method
Showing 1-1 of 1
Compare
Product Number
Description
Pricing