
Nateglinide impurity A

Synonym(s):
trans-4-Isopropylcyclohexanecarboxylic acid
Empirical Formula (Hill Notation):
C10H18O2
CAS Number:
Molecular Weight:
170.25
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing