
HMX solution

Empirical Formula (Hill Notation):
C4H8N8O8
CAS Number:
Molecular Weight:
296.16
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing