Skip to Content
Merck
γ-Hexalactone analytical standard

γ-Hexalactone

γ-Hexalactone analytical standard
Synonym(s):
γ-Caprolactone, γ-Ethyl-γ-butyrolactone
Empirical Formula (Hill Notation):
C6H10O2
CAS Number:
Molecular Weight:
114.14
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Gene Alias
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing